Product Details

SNP ID
rs145402588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27451610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTAGATGGGAGCTCTGGCTAAAG[A/G]TCTTTTCACATTTATTACACTGGTA
Phenotype
MIM: 602277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF184 PubMed Links

Gene Details

Gene
ZNF184
Gene Name
zinc finger protein 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318891.1 2234 Missense Mutation ACC,ATC T650I NP_001305820.1
NM_001318892.1 2234 Missense Mutation ACC,ATC T650I NP_001305821.1
NM_001318893.1 2234 Missense Mutation ACC,ATC T574I NP_001305822.1
NM_007149.2 2234 Missense Mutation ACC,ATC T650I NP_009080.2
XM_011514861.2 2234 Missense Mutation ACC,ATC T574I XP_011513163.1

View Full Product Details