Product Details

SNP ID
rs145437840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:16130652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTATGGCTAAACCTGAGAAACCG[C/G]ATCTCCCAGCAGATGGATGGGCTAG
Phenotype
MIM: 610082
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYLIP PubMed Links

Gene Details

Gene
MYLIP
Gene Name
myosin regulatory light chain interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013262.3 420 Silent Mutation CGC,CGG R61R NP_037394.2
XM_005249032.2 420 Silent Mutation CGC,CGG R61R XP_005249089.1
XM_005249033.2 420 Intron XP_005249090.1
XM_017010789.1 420 Silent Mutation CGC,CGG R61R XP_016866278.1

View Full Product Details