Product Details

SNP ID
rs145607851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107871353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTAAAGCTGAACAAAGAAAAGTC[A/G]CTCATTTACAGACTGTGTTTCCTGC
Phenotype
MIM: 608648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC63 PubMed Links

Gene Details

Gene
SEC63
Gene Name
SEC63 homolog, protein translocation regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007214.4 2652 UTR 3 NP_009145.1
XM_011535399.2 2652 UTR 3 XP_011533701.1
XM_017010218.1 2652 UTR 3 XP_016865707.1

View Full Product Details