Product Details

SNP ID
rs145784294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49731204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTTATCTGGGCAACTGGCACAA[A/G]GTGCTCCTTGTTCATAAGGGACATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRISP3 PubMed Links

Gene Details

Gene
CRISP3
Gene Name
cysteine rich secretory protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190986.1 696 Missense Mutation CCT,CTT P213L NP_001177915.1
NM_006061.2 696 Missense Mutation CCT,CTT P203L NP_006052.2

View Full Product Details