Product Details

SNP ID
rs146279104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42184912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACCCACTTGTCCCGACGGGCA[C/T]CTTCAACAAACTCGTTCAGAGACAG
Phenotype
MIM: 600364 MIM: 602275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCA1A PubMed Links

Gene Details

Gene
GUCA1A
Gene Name
guanylate cyclase activator 1A
There are no transcripts associated with this gene.

Gene
GUCA1B
Gene Name
guanylate cyclase activator 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002098.5 642 Missense Mutation GAT,GGT D169G NP_002089.4

View Full Product Details