Product Details

SNP ID
rs146500509
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:113940827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTGACATAATAACTCACATCA[A/G]TTTTAAATACTATCACATAAAGCAT
Phenotype
MIM: 605164
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HDAC2 PubMed Links

Gene Details

Gene
HDAC2
Gene Name
histone deacetylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001527.3 1959 UTR 3 NP_001518.3
XM_011535788.1 1959 UTR 3 XP_011534090.1
XM_017010799.1 1959 UTR 3 XP_016866288.1

View Full Product Details