Product Details

SNP ID
rs146842669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137198281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTATCAAAACCATTTCTGGAG[G/T]GATCACTCTCAGAACAATTTCTGGA
Phenotype
MIM: 107470
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IFNGR1 PubMed Links

Gene Details

Gene
IFNGR1
Gene Name
interferon gamma receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000416.2 1394 Missense Mutation NP_000407.1
XM_006715470.2 1394 Missense Mutation XP_006715533.1
XM_006715471.2 1394 Missense Mutation XP_006715534.1
XM_011535793.2 1394 Missense Mutation XP_011534095.1
XM_011535794.1 1394 Missense Mutation XP_011534096.1
XM_017010827.1 1394 Missense Mutation XP_016866316.1

View Full Product Details