Product Details

SNP ID
rs146961852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28259781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAATTGGGAGCGCCTGAAGTGT[G/T]GGGGCCGTCTCCAAAGTTCCCTCAG
Phenotype
MIM: 611643 MIM: 616474
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NKAPL PubMed Links

Gene Details

Gene
NKAPL
Gene Name
NFKB activating protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007531.2 485 Missense Mutation TGG,TTG W137L NP_001007532.1
Gene
ZKSCAN4
Gene Name
zinc finger with KRAB and SCAN domains 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304506.1 485 Intron NP_001291435.1
NM_019110.4 485 Intron NP_061983.2
XM_005249095.3 485 Intron XP_005249152.1
XM_005249097.3 485 Intron XP_005249154.1
XM_005249098.3 485 Intron XP_005249155.1
XM_005249100.4 485 Intron XP_005249157.2
XM_006715095.3 485 Intron XP_006715158.1
XM_011514588.2 485 Intron XP_011512890.1
XM_011514589.2 485 Intron XP_011512891.1
XM_017010844.1 485 Intron XP_016866333.1
Gene
ZSCAN26
Gene Name
zinc finger and SCAN domain containing 26
There are no transcripts associated with this gene.

View Full Product Details