Product Details
- SNP ID
-
rs147099303
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:41937291 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCTTTTTGACCAAGGCCTGTCGG[G/T]CACGGGGCAGAGAGAGCCGGTGCAG
- Phenotype
-
MIM: 603871
MIM: 123834
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BYSL
PubMed Links
Gene Details
- Gene
- BYSL
- Gene Name
- bystin like
There are no transcripts associated with this gene.
- Gene
- CCND3
- Gene Name
- cyclin D3
View Full Product Details