Product Details

SNP ID
rs147401311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29487059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAATCTGTTATGAGGGGTGCCA[C/T]GCTCAGGGGTAGGGCCCAGAGTAGG
Phenotype
MIM: 607235
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAS1L PubMed Links

Gene Details

Gene
MAS1L
Gene Name
MAS1 proto-oncogene like, G protein-coupled receptor
There are no transcripts associated with this gene.

View Full Product Details