Product Details

SNP ID
rs148067109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACGGCCCTGCTGCTGCTCCTAGC[C/G]TCGCAGGTCCTGTCTCCGGGAAGCT
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 408 Silent Mutation GCC,GCG A20A NP_001025034.1
NM_001317342.1 408 Silent Mutation GCC,GCG A39A NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 408 Intron NP_001189479.1
NM_182552.4 408 Intron NP_872358.4
XM_011535682.2 408 Intron XP_011533984.1
XM_011535684.2 408 Intron XP_011533986.1
XM_011535685.2 408 Intron XP_011533987.1
XM_011535687.2 408 Intron XP_011533989.1
XM_011535688.2 408 Intron XP_011533990.1
XM_011535689.2 408 Intron XP_011533991.1
XM_011535690.2 408 Intron XP_011533992.1
XM_011535691.2 408 Intron XP_011533993.1
XM_011535692.2 408 Intron XP_011533994.1
XM_011535693.2 408 Intron XP_011533995.1
XM_011535694.2 408 Intron XP_011533996.1
XM_011535696.2 408 Intron XP_011533998.1
XM_011535697.2 408 Intron XP_011533999.1
XM_017010658.1 408 Intron XP_016866147.1
XM_017010659.1 408 Intron XP_016866148.1
XM_017010660.1 408 Intron XP_016866149.1
XM_017010661.1 408 Intron XP_016866150.1
XM_017010662.1 408 Intron XP_016866151.1
XM_017010663.1 408 Intron XP_016866152.1
XM_017010664.1 408 Intron XP_016866153.1
XM_017010665.1 408 Intron XP_016866154.1
XM_017010666.1 408 Intron XP_016866155.1
XM_017010667.1 408 Intron XP_016866156.1
XM_017010668.1 408 Intron XP_016866157.1
XM_017010669.1 408 Intron XP_016866158.1
XM_017010670.1 408 Intron XP_016866159.1
XM_017010671.1 408 Intron XP_016866160.1
XM_017010672.1 408 Intron XP_016866161.1
XM_017010673.1 408 Intron XP_016866162.1
XM_017010674.1 408 Intron XP_016866163.1
XM_017010675.1 408 Intron XP_016866164.1

View Full Product Details