Product Details

SNP ID
rs148280839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149818943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATGCCTCATCAGTTTTTATTTT[A/G]TTGGTTATGGCTATAGTTGACATCT
Phenotype
MIM: 176851
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRP11 PubMed Links

Gene Details

Gene
LRP11
Gene Name
LDL receptor related protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032832.5 3433 UTR 3 NP_116221.3
XM_011536194.2 3433 UTR 3 XP_011534496.1
XM_017011380.1 3433 Intron XP_016866869.1
Gene
PCMT1
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase
There are no transcripts associated with this gene.

View Full Product Details