Product Details
- SNP ID
-
rs148433943
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:30186260 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCCAACGATTCCTCATCTTCGTC[C/T]GTTTCCCAGTCGTCATATCCATCCC
- Phenotype
-
MIM: 600830
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRIM26
PubMed Links
Gene Details
- Gene
- TRIM26
- Gene Name
- tripartite motif containing 26
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242783.1 |
1366 |
Silent Mutation |
ACA,ACG |
T412T |
NP_001229712.1 |
NM_003449.4 |
1366 |
Silent Mutation |
ACA,ACG |
T412T |
NP_003440.1 |
XM_005249374.2 |
1366 |
Silent Mutation |
ACA,ACG |
T412T |
XP_005249431.1 |
XM_005249375.2 |
1366 |
Silent Mutation |
ACA,ACG |
T412T |
XP_005249432.1 |
XM_005249376.2 |
1366 |
Silent Mutation |
ACA,ACG |
T412T |
XP_005249433.1 |
XM_005249377.2 |
1366 |
Silent Mutation |
ACA,ACG |
T412T |
XP_005249434.1 |
XM_005249378.2 |
1366 |
Silent Mutation |
ACA,ACG |
T412T |
XP_005249435.1 |
XM_006715180.2 |
1366 |
Silent Mutation |
ACA,ACG |
T412T |
XP_006715243.1 |
XM_011514859.1 |
1366 |
Silent Mutation |
ACA,ACG |
T335T |
XP_011513161.1 |
XM_017011263.1 |
1366 |
Silent Mutation |
ACA,ACG |
T335T |
XP_016866752.1 |
View Full Product Details