Product Details

SNP ID
rs148473967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:57191413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTGCATCTTTTGACTTAACAAT[C/T]TATCTTTGACGTTTGGAATCAGCAA
Phenotype
MIM: 603882 MIM: 606144
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAG2 PubMed Links

Gene Details

Gene
BAG2
Gene Name
BCL2 associated athanogene 2
There are no transcripts associated with this gene.

Gene
RAB23
Gene Name
RAB23, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278666.1 Intron NP_001265595.1
NM_001278667.1 Intron NP_001265596.1
NM_001278668.1 Intron NP_001265597.1
NM_016277.4 Intron NP_057361.3
NM_183227.2 Intron NP_899050.1

View Full Product Details