Product Details

SNP ID
rs149235880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2887465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATTCAAGAGCCAATAAATGCA[C/T]AGAACACTTTCACCACTAATAAAAG
Phenotype
MIM: 601799
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927730 PubMed Links

Gene Details

Gene
LOC101927730
Gene Name
uncharacterized LOC101927730
There are no transcripts associated with this gene.

Gene
SERPINB9
Gene Name
serpin family B member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004155.5 4101 UTR 3 NP_004146.1
XM_005249184.4 4101 UTR 3 XP_005249241.1
XM_011514678.2 4101 UTR 3 XP_011512980.1
XM_017010942.1 4101 UTR 3 XP_016866431.1
XM_017010943.1 4101 UTR 3 XP_016866432.1

View Full Product Details