Product Details

SNP ID
rs149460386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49435621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTGCTGGGCCACTTCACGAGG[A/G]GTCTAAACAGTCAGAAAGTAAAGAT
Phenotype
MIM: 609058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUT PubMed Links

Gene Details

Gene
MUT
Gene Name
methylmalonyl-CoA mutase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000255.3 2224 Silent Mutation ACC,ACT T653T NP_000246.2
XM_005249143.3 2224 Silent Mutation ACC,ACT T653T XP_005249200.1

View Full Product Details