Product Details

SNP ID
rs149461273
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32188140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAGCAGGTTCATGACATGGGTC[A/G]TGAACTCATTACATGCCTGTAGTGG
Phenotype
MIM: 600214 MIM: 164951 MIM: 176311 MIM: 602677
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AGER PubMed Links

Gene Details

Gene
AGER
Gene Name
advanced glycosylation end product-specific receptor
There are no transcripts associated with this gene.

Gene
GPSM3
Gene Name
G-protein signaling modulator 3
There are no transcripts associated with this gene.

Gene
MIR6833
Gene Name
microRNA 6833
There are no transcripts associated with this gene.

Gene
NOTCH4
Gene Name
notch 4
There are no transcripts associated with this gene.

Gene
PBX2
Gene Name
PBX homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002586.4 Intron NP_002577.2
Gene
RNF5
Gene Name
ring finger protein 5
There are no transcripts associated with this gene.

View Full Product Details