Product Details
- SNP ID
-
rs150031834
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:107869865 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCAGAGAAGCCAGCTGTGTGAAT[C/G]AACTTATAAAGCCTCTTTAAGGCAA
- Phenotype
-
MIM: 608648
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SEC63
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs55885857] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEC63
- Gene Name
- SEC63 homolog, protein translocation regulator
View Full Product Details