Product Details

SNP ID
rs150069419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26031929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCTCGCGCAGAGCCACAGTGCC[C/T]GGGCGGTAACGGTGAGGCTTTTTCA
Phenotype
MIM: 602795 MIM: 602819 MIM: 602822 MIM: 602829
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AB PubMed Links

Gene Details

Gene
HIST1H2AB
Gene Name
histone cluster 1, H2ab
There are no transcripts associated with this gene.

Gene
HIST1H3B
Gene Name
histone cluster 1, H3b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003537.3 Intron NP_003528.1
Gene
HIST1H4A
Gene Name
histone cluster 1, H4a
There are no transcripts associated with this gene.

Gene
HIST1H4B
Gene Name
histone cluster 1, H4b
There are no transcripts associated with this gene.

View Full Product Details