Product Details

SNP ID
rs150294928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2948588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTGCGCAGGACACTCTCCATGT[C/T]GTAGCTTTCCTCTAGTTTAAACCGC
Phenotype
MIM: 173321
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINB6 PubMed Links

Gene Details

Gene
SERPINB6
Gene Name
serpin family B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195291.2 1425 Missense Mutation NP_001182220.2
NM_001271822.1 1425 Missense Mutation NP_001258751.1
NM_001271823.1 1425 Missense Mutation NP_001258752.1
NM_001271824.1 1425 Missense Mutation NP_001258753.1
NM_001271825.1 1425 Missense Mutation NP_001258754.1
NM_001297699.1 1425 Missense Mutation NP_001284628.1
NM_001297700.1 1425 Missense Mutation NP_001284629.1
NM_004568.5 1425 Missense Mutation NP_004559.4
XM_011514672.1 1425 Missense Mutation XP_011512974.1
XM_011514673.1 1425 Missense Mutation XP_011512975.1
XM_011514674.2 1425 Missense Mutation XP_011512976.1
XM_011514676.2 1425 Missense Mutation XP_011512978.1
XM_017010940.1 1425 Missense Mutation XP_016866429.1
XM_017010941.1 1425 Missense Mutation XP_016866430.1

View Full Product Details