Product Details

SNP ID
rs150634482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26156513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGCGTCTGGGCCCCCGGTGTC[C/T]GAGCTCATTACTAAAGCTGTTGCCG
Phenotype
MIM: 142220 MIM: 602799
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H1E PubMed Links

Gene Details

Gene
HIST1H1E
Gene Name
histone cluster 1, H1e
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005321.2 183 Silent Mutation TCC,TCT S41S NP_005312.1
Gene
HIST1H2BD
Gene Name
histone cluster 1, H2bd
There are no transcripts associated with this gene.

View Full Product Details