Product Details

SNP ID
rs151039809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33319663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTTCCTCCAGGGTCAGCTCCT[C/T]AGGCTGTTCTCCATTGCTTTCAGCA
Phenotype
MIM: 603186 MIM: 601962 MIM: 611439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAXX PubMed Links

Gene Details

Gene
DAXX
Gene Name
death domain associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141969.1 711 Missense Mutation AAG,GAG K553E NP_001135441.1
NM_001141970.1 711 Missense Mutation AAG,GAG K565E NP_001135442.1
NM_001254717.1 711 Missense Mutation AAG,GAG K478E NP_001241646.1
NM_001350.4 711 Missense Mutation AAG,GAG K553E NP_001341.1
XM_005248860.3 711 Missense Mutation AAG,GAG K185E XP_005248917.1
Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
There are no transcripts associated with this gene.

Gene
ZBTB22
Gene Name
zinc finger and BTB domain containing 22
There are no transcripts associated with this gene.

View Full Product Details