Product Details

SNP ID
rs151065034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3072276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATTGTGGAAAATTTGGAAAACA[C/T]AGTAAAAGTGTAAAGAAGAAAGTAA
Phenotype
MIM: 603453
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RIPK1 PubMed Links

Gene Details

Gene
RIPK1
Gene Name
receptor interacting serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249457.3 Intron XP_005249514.2
XM_006715237.3 Intron XP_006715300.2
XM_017011403.1 Intron XP_016866892.1
XM_017011404.1 Intron XP_016866893.1
XM_017011405.1 Intron XP_016866894.1

View Full Product Details