Product Details

SNP ID
rs151083087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149942103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCCGCCGCTACCAAGATCCTT[C/T]TGTGCCTCCCGCTTCTGCTCCTGCT
Phenotype
MIM: 605698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105378052 PubMed Links

Gene Details

Gene
LOC105378052
Gene Name
uncharacterized LOC105378052
There are no transcripts associated with this gene.

Gene
ULBP2
Gene Name
UL16 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025217.3 Intron NP_079493.1
XM_017011321.1 Intron XP_016866810.1

View Full Product Details