Product Details

SNP ID
rs137974406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:50321269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCCTTAAGCGTTTACTTTGTTT[C/T]GTAAACAAAAAAGCACAAAAACAAA
Phenotype
MIM: 603023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKZF1 PubMed Links

Gene Details

Gene
IKZF1
Gene Name
IKAROS family zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220765.2 Intron NP_001207694.1
NM_001220767.2 Intron NP_001207696.1
NM_001220768.2 Intron NP_001207697.1
NM_001220770.2 Intron NP_001207699.1
NM_001220771.2 Intron NP_001207700.1
NM_001291837.1 Intron NP_001278766.1
NM_001291838.1 Intron NP_001278767.1
NM_001291839.1 Intron NP_001278768.1
NM_001291840.1 Intron NP_001278769.1
NM_001291841.1 Intron NP_001278770.1
NM_001291842.1 Intron NP_001278771.1
NM_001291843.1 Intron NP_001278772.1
NM_001291844.1 Intron NP_001278773.1
NM_001291845.1 Intron NP_001278774.1
NM_001291846.1 Intron NP_001278775.1
NM_001291847.1 Intron NP_001278776.1
NM_006060.5 Intron NP_006051.1
XM_011515058.2 Intron XP_011513360.1
XM_011515059.2 Intron XP_011513361.1
XM_011515060.2 Intron XP_011513362.1
XM_011515061.2 Intron XP_011513363.1
XM_011515062.2 Intron XP_011513364.1
XM_011515063.2 Intron XP_011513365.1
XM_011515064.2 Intron XP_011513366.1
XM_011515065.2 Intron XP_011513367.1
XM_011515066.2 Intron XP_011513368.1
XM_011515067.2 Intron XP_011513369.1
XM_011515068.2 Intron XP_011513370.1
XM_011515069.2 Intron XP_011513371.1
XM_011515070.2 Intron XP_011513372.1
XM_011515071.2 Intron XP_011513373.1
XM_011515072.2 Intron XP_011513374.1
XM_011515073.2 Intron XP_011513375.1
XM_011515074.2 Intron XP_011513376.1
XM_011515075.2 Intron XP_011513377.1
XM_011515076.2 Intron XP_011513378.1
XM_011515077.2 Intron XP_011513379.1
XM_011515078.2 Intron XP_011513380.1
XM_017011667.1 Intron XP_016867156.1
XM_017011668.1 Intron XP_016867157.1
XM_017011669.1 Intron XP_016867158.1
XM_017011670.1 Intron XP_016867159.1

View Full Product Details