Product Details

SNP ID
rs138919994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101234309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGTGGAGGGCCCCTGCGGTGGC[C/T]GCCAGCTTGGCTTTCCTGGAGAGCT
Phenotype
MIM: 615778 MIM: 609003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN15 PubMed Links

Gene Details

Gene
CLDN15
Gene Name
claudin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185080.1 716 Silent Mutation GCA,GCG A117A NP_001172009.1
NM_014343.2 716 Silent Mutation GCA,GCG A117A NP_055158.1
Gene
FIS1
Gene Name
fission, mitochondrial 1
There are no transcripts associated with this gene.

View Full Product Details