Product Details
- SNP ID
-
rs140516632
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149103941 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGTACTGTTGAGTGAAACTTTTGC[C/G]GCACATCACACAAGAGAATGGCTTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF425
PubMed Links
Gene Details
- Gene
- ZNF425
- Gene Name
- zinc finger protein 425
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001661.2 |
2063 |
Missense Mutation |
CGC,GGC |
R644G |
NP_001001661.1 |
View Full Product Details