Product Details

SNP ID
rs140836534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:88209998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATACCTACCCATTGTTGTCAGGG[C/T]CTTCTGCAATTCTTGTGGGTCTACT
Phenotype
MIM: 603709 MIM: 182520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM22 PubMed Links

Gene Details

Gene
ADAM22
Gene Name
ADAM metallopeptidase domain 22
There are no transcripts associated with this gene.

Gene
LOC102723885
Gene Name
uncharacterized LOC102723885
There are no transcripts associated with this gene.

Gene
SRI
Gene Name
sorcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256891.1 440 Missense Mutation ACC,GCC T128A NP_001243820.1
NM_001256892.1 440 Missense Mutation ACC,GCC T113A NP_001243821.1
NM_003130.3 440 Missense Mutation ACC,GCC T128A NP_003121.1
NM_198901.1 440 Missense Mutation ACC,GCC T113A NP_944490.1
XM_011516528.1 440 Missense Mutation ACC,GCC T85A XP_011514830.1

View Full Product Details