Product Details

SNP ID
rs140883968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:116526641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACGACGCGCACACCAAGGAGAT[C/T]GACCTGGTCAACCGCGACCCTAAAC
Phenotype
MIM: 601047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAV1 PubMed Links

Gene Details

Gene
CAV1
Gene Name
caveolin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172895.1 425 Silent Mutation ATC,ATT I18I NP_001166366.1
NM_001172896.1 425 Silent Mutation ATC,ATT I18I NP_001166367.1
NM_001172897.1 425 Silent Mutation ATC,ATT I18I NP_001166368.1
NM_001753.4 425 Silent Mutation ATC,ATT I49I NP_001744.2

View Full Product Details