Product Details

SNP ID
rs141212035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:155502622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGCCACATTTTGAATTTGTTG[C/G]CATGTATTTCCTTAAATTTATAGGC
Phenotype
MIM: 612493
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNPY1 PubMed Links

Gene Details

Gene
CNPY1
Gene Name
canopy FGF signaling regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103176.1 870 UTR 3 NP_001096646.1

View Full Product Details