Product Details

SNP ID
rs141528720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6622537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGGTGAGCGGCCACACCGCTGC[G/T]GCGAGTGCGGCAAGGGCTTCTCGCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF853 PubMed Links

Gene Details

Gene
ZNF853
Gene Name
zinc finger protein 853
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017560.1 1987 Missense Mutation GGC,TGC G516C NP_060030.1
XM_011515438.2 1987 Missense Mutation GGC,TGC G542C XP_011513740.1
XM_011515439.2 1987 Missense Mutation GGC,TGC G527C XP_011513741.1
XM_017012371.1 1987 Missense Mutation GGC,TGC G501C XP_016867860.1

View Full Product Details