Product Details
- SNP ID
-
rs141539487
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:107571212 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTACACACCAATGATTGTTGCC[A/G]CTGATTTTGTCAAATCTATAAAAGC
- Phenotype
-
MIM: 606821
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BCAP29
PubMed Links
Gene Details
- Gene
- BCAP29
- Gene Name
- B-cell receptor-associated protein 29
There are no transcripts associated with this gene.
- Gene
- COG5
- Gene Name
- component of oligomeric golgi complex 5
There are no transcripts associated with this gene.
- Gene
- DUS4L
- Gene Name
- dihydrouridine synthase 4 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001270419.1 |
291 |
Missense Mutation |
ACT,GCT |
T62A |
NP_001257348.1 |
NM_181581.2 |
291 |
Missense Mutation |
ACT,GCT |
T62A |
NP_853559.1 |
XM_005250125.4 |
291 |
Missense Mutation |
ACT,GCT |
T62A |
XP_005250182.1 |
XM_006715844.2 |
291 |
Intron |
|
|
XP_006715907.1 |
XM_006715845.3 |
291 |
Intron |
|
|
XP_006715908.1 |
XM_011515766.2 |
291 |
Missense Mutation |
ACT,GCT |
T62A |
XP_011514068.1 |
XM_011515767.2 |
291 |
Missense Mutation |
ACT,GCT |
T62A |
XP_011514069.1 |
XM_017011708.1 |
291 |
Missense Mutation |
ACT,GCT |
T62A |
XP_016867197.1 |
XM_017011709.1 |
291 |
Missense Mutation |
ACT,GCT |
T58A |
XP_016867198.1 |
XM_017011710.1 |
291 |
Missense Mutation |
ACT,GCT |
T58A |
XP_016867199.1 |
XM_017011711.1 |
291 |
Missense Mutation |
ACT,GCT |
T58A |
XP_016867200.1 |
XM_017011712.1 |
291 |
Missense Mutation |
ACT,GCT |
T58A |
XP_016867201.1 |
XM_017011713.1 |
291 |
Missense Mutation |
ACT,GCT |
T58A |
XP_016867202.1 |
View Full Product Details