Product Details

SNP ID
rs141691628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:84997671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTCTCAACATAGAGCCACAGAT[C/G]TTGCATTAAGTTGCCCTCTAATCAC
Phenotype
MIM: 609907
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SEMA3D PubMed Links

Gene Details

Gene
SEMA3D
Gene Name
semaphorin 3D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152754.2 4642 UTR 3 NP_689967.2
XM_011515960.1 4642 UTR 3 XP_011514262.1
XM_011515961.2 4642 UTR 3 XP_011514263.1
XM_017011873.1 4642 UTR 3 XP_016867362.1

View Full Product Details