Product Details

SNP ID
rs141907752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75931913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATTCAGTTTGCCCTCCCTGCTGG[A/G]GCACAGGTGTCATGGCCTCCCTTAA
Phenotype
MIM: 124015
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POR PubMed Links

Gene Details

Gene
POR
Gene Name
cytochrome p450 oxidoreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000941.2 Intron NP_000932.3

View Full Product Details