Product Details
- SNP ID
-
rs142567786
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:150388348 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGAGAGTGCTTTATGGGATGGGA[G/T]GGGGGCTGGGGACCCTGAGTTTTGT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF775
PubMed Links
Gene Details
- Gene
- ZNF775
- Gene Name
- zinc finger protein 775
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173680.3 |
|
Intron |
|
|
NP_775951.2 |
View Full Product Details