Product Details

SNP ID
rs142778277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99890957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAATATATTGAAATGTTAGTA[A/G]TGCATAACTTTAATTTTTATACTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM4 PubMed Links

Gene Details

Gene
TRIM4
Gene Name
tripartite motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033017.3 2830 UTR 3 NP_148977.2
NM_033091.2 2830 UTR 3 NP_149082.1

View Full Product Details