Product Details
- SNP ID
-
rs142846778
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:81950430 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGTGCTGCCAGATACCAGCCAAA[A/G]TAGTAGAAACTGGATTCCAATGATA
- Phenotype
-
MIM: 114204
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CACNA2D1
PubMed Links
Gene Details
- Gene
- CACNA2D1
- Gene Name
- calcium voltage-gated channel auxiliary subunit alpha2delta 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000722.3 |
3713 |
Missense Mutation |
CTT,TTT |
L1080F |
NP_000713.2 |
NM_001302890.1 |
3713 |
Intron |
|
|
NP_001289819.1 |
XM_005250570.2 |
3713 |
Missense Mutation |
CTT,TTT |
L1092F |
XP_005250627.1 |
XM_005250572.2 |
3713 |
Missense Mutation |
CTT,TTT |
L1075F |
XP_005250629.1 |
XM_005250573.2 |
3713 |
Missense Mutation |
CTT,TTT |
L1073F |
XP_005250630.1 |
XM_005250574.2 |
3713 |
Missense Mutation |
CTT,TTT |
L1068F |
XP_005250631.1 |
XM_006716118.2 |
3713 |
Missense Mutation |
CTT,TTT |
L1099F |
XP_006716181.1 |
XM_006716119.3 |
3713 |
Missense Mutation |
CTT,TTT |
L1074F |
XP_006716182.1 |
XM_006716120.3 |
3713 |
Missense Mutation |
CTT,TTT |
L1060F |
XP_006716183.1 |
XM_006716121.3 |
3713 |
Missense Mutation |
CTT,TTT |
L569F |
XP_006716184.1 |
XM_011516570.2 |
3713 |
Intron |
|
|
XP_011514872.1 |
XM_011516571.2 |
3713 |
Missense Mutation |
CTT,TTT |
L1094F |
XP_011514873.1 |
XM_011516572.2 |
3713 |
Missense Mutation |
CTT,TTT |
L1087F |
XP_011514874.1 |
XM_017012588.1 |
3713 |
Missense Mutation |
CTT,TTT |
L1041F |
XP_016868077.1 |
View Full Product Details