Product Details

SNP ID
rs142899940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122076539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATATATAATGCCTTCTGCTTTAA[A/T]TCGCTCCAATATTGGTCCATAGATC
Phenotype
MIM: 605113
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AASS PubMed Links

Gene Details

Gene
AASS
Gene Name
aminoadipate-semialdehyde synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005763.3 2876 Missense Mutation ATT,TTT I911F NP_005754.2
XM_011515725.2 2876 Missense Mutation ATT,TTT I879F XP_011514027.1

View Full Product Details