Product Details

SNP ID
rs143213043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123542218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGCCAGATTTAGTTCATGTTC[A/G]GCCTGTTAATACAAATTTTTTAAAA
Phenotype
MIM: 601677
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQUB PubMed Links

Gene Details

Gene
IQUB
Gene Name
IQ motif and ubiquitin domain containing
There are no transcripts associated with this gene.

Gene
NDUFA5
Gene Name
NADH:ubiquinone oxidoreductase subunit A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282419.2 555 Silent Mutation GCC,GCT A69A NP_001269348.1
NM_001282420.2 555 Silent Mutation GCC,GCT A84A NP_001269349.1
NM_001282421.2 555 Silent Mutation GCC,GCT A81A NP_001269350.1
NM_001282422.2 555 Silent Mutation GCC,GCT A45A NP_001269351.1
NM_001291304.1 555 Silent Mutation GCC,GCT A150A NP_001278233.1
NM_005000.4 555 Silent Mutation GCC,GCT A84A NP_004991.1

View Full Product Details