Product Details

SNP ID
rs143309023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75811838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGATATCTCTGGACAGGGCC[C/T]TTGACAGCCACTGCCCTGGCCCTAG
Phenotype
MIM: 602495
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL24 PubMed Links

Gene Details

Gene
CCL24
Gene Name
C-C motif chemokine ligand 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002991.2 585 Silent Mutation AAA,AAG K106K NP_002982.2
XM_011516459.2 585 Silent Mutation AAA,AAG K106K XP_011514761.1
XM_011516460.2 585 Silent Mutation AAA,AAG K106K XP_011514762.1

View Full Product Details