Product Details
- SNP ID
-
rs143495981
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:65967904 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCAAACAGATCACATCCACATACG[C/G]AGCCTAGGACCAGAGCAGCAGAGCC
- Phenotype
-
MIM: 611499
MIM: 608838
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GUSB
PubMed Links
Gene Details
- Gene
- GUSB
- Gene Name
- glucuronidase beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000181.3 |
1355 |
Missense Mutation |
CCG,GCG |
P494A |
NP_000172.2 |
NM_001284290.1 |
1355 |
Missense Mutation |
CCG,GCG |
P348A |
NP_001271219.1 |
NM_001293104.1 |
1355 |
Missense Mutation |
CCG,GCG |
P304A |
NP_001280033.1 |
NM_001293105.1 |
1355 |
Missense Mutation |
CCG,GCG |
P275A |
NP_001280034.1 |
XM_005250297.3 |
1355 |
Missense Mutation |
CCG,GCG |
P443A |
XP_005250354.1 |
XM_011516113.1 |
1355 |
Missense Mutation |
CCG,GCG |
P327A |
XP_011514415.1 |
XM_011516114.1 |
1355 |
Missense Mutation |
CCG,GCG |
P270A |
XP_011514416.1 |
XM_017012091.1 |
1355 |
Missense Mutation |
CCG,GCG |
P276A |
XP_016867580.1 |
XM_017012092.1 |
1355 |
Missense Mutation |
CCG,GCG |
P253A |
XP_016867581.1 |
XM_017012093.1 |
1355 |
Missense Mutation |
CCG,GCG |
P219A |
XP_016867582.1 |
- Gene
- VKORC1L1
- Gene Name
- vitamin K epoxide reductase complex subunit 1 like 1
There are no transcripts associated with this gene.
View Full Product Details