Product Details

SNP ID
rs143631985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:81950402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTTTTTAGAAGGTCATAACAGG[C/T]GGTGTGTGCTGCCAGATACCAGCCA
Phenotype
MIM: 114204
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNA2D1 PubMed Links

Gene Details

Gene
CACNA2D1
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000722.3 3741 Missense Mutation NP_000713.2
NM_001302890.1 3741 Intron NP_001289819.1
XM_005250570.2 3741 Missense Mutation XP_005250627.1
XM_005250572.2 3741 Missense Mutation XP_005250629.1
XM_005250573.2 3741 Missense Mutation XP_005250630.1
XM_005250574.2 3741 Missense Mutation XP_005250631.1
XM_006716118.2 3741 Missense Mutation XP_006716181.1
XM_006716119.3 3741 Missense Mutation XP_006716182.1
XM_006716120.3 3741 Missense Mutation XP_006716183.1
XM_006716121.3 3741 Missense Mutation XP_006716184.1
XM_011516570.2 3741 Intron XP_011514872.1
XM_011516571.2 3741 Missense Mutation XP_011514873.1
XM_011516572.2 3741 Missense Mutation XP_011514874.1
XM_017012588.1 3741 Missense Mutation XP_016868077.1

View Full Product Details