Product Details

SNP ID
rs144031260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100723103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGGGAAAGCTGAAGCTGTACAC[A/C]GGGGAGGCCTGCAGGACAGGGGACA
Phenotype
MIM: 133170
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EPO PubMed Links

Gene Details

Gene
EPO
Gene Name
erythropoietin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000799.2 733 Silent Mutation ACA,ACC T184T NP_000790.2

View Full Product Details