Product Details

SNP ID
rs144148146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2235375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAGCCTGTCATGATCGAGGTCC[C/T]GGAACCCTGTGGCATTGGGCGCCAT
Phenotype
MIM: 602686 MIM: 606906 MIM: 600312
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAD1L1 PubMed Links

Gene Details

Gene
MAD1L1
Gene Name
MAD1 mitotic arrest deficient like 1
There are no transcripts associated with this gene.

Gene
MRM2
Gene Name
mitochondrial rRNA methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013393.1 517 Missense Mutation CAG,CGG Q163R NP_037525.1
Gene
NUDT1
Gene Name
nudix hydrolase 1
There are no transcripts associated with this gene.

View Full Product Details