Product Details

SNP ID
rs144181064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:14147134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCATAGATACAAAAAATCAATG[C/T]GTGTGTTACGTAACATTATGTCAGT
Phenotype
MIM: 604070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGKB PubMed Links

Gene Details

Gene
DGKB
Gene Name
diacylglycerol kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004080.2 4599 UTR 3 NP_004071.1
NM_145695.2 4599 Intron NP_663733.1
XM_005249628.2 4599 Intron XP_005249685.1
XM_005249629.2 4599 Intron XP_005249686.1
XM_005249630.2 4599 Intron XP_005249687.1
XM_005249631.3 4599 Intron XP_005249688.1
XM_011515153.2 4599 Intron XP_011513455.1
XM_011515154.2 4599 Intron XP_011513456.1
XM_011515156.2 4599 Intron XP_011513458.1
XM_011515157.2 4599 Intron XP_011513459.1
XM_011515158.2 4599 Intron XP_011513460.1
XM_017011783.1 4599 Intron XP_016867272.1
XM_017011784.1 4599 Intron XP_016867273.1
XM_017011785.1 4599 Intron XP_016867274.1
XM_017011786.1 4599 Intron XP_016867275.1
XM_017011787.1 4599 Intron XP_016867276.1
XM_017011788.1 4599 Intron XP_016867277.1
XM_017011789.1 4599 Intron XP_016867278.1
XM_017011790.1 4599 Intron XP_016867279.1
XM_017011791.1 4599 Intron XP_016867280.1
XM_017011792.1 4599 Intron XP_016867281.1

View Full Product Details