Product Details

SNP ID
rs144470429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27526307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACATCATCCTGTAGATCTGATG[C/G]GCCAGACTGCCAAGAAGGATTGGGC
Phenotype
MIM: 608475
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIBADH PubMed Links

Gene Details

Gene
HIBADH
Gene Name
3-hydroxyisobutyrate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152740.3 1161 Silent Mutation GCC,GCG A306A NP_689953.1
XM_017011719.1 1161 Silent Mutation GCC,GCG A205A XP_016867208.1

View Full Product Details