Product Details

SNP ID
rs145023433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141010627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCGGAAGGCATATTCTGAAAGGG[A/T]GGACATTTCTTGAGTGGCAAGGAGT
Phenotype
MIM: 611993
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPS33 PubMed Links

Gene Details

Gene
MRPS33
Gene Name
mitochondrial ribosomal protein S33
There are no transcripts associated with this gene.

View Full Product Details