Product Details

SNP ID
rs145354319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:49775441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCGGGACGTGGTAGGGGATGCC[C/T]AGCTCCACTGCGATGGCAGTTGGCG
Phenotype
MIM: 611108
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VWC2 PubMed Links

Gene Details

Gene
VWC2
Gene Name
von Willebrand factor C domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198570.4 562 Silent Mutation CCC,CCT P2P NP_940972.2

View Full Product Details