Product Details

SNP ID
rs145875827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:20785652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTGCAGCTGGCGGAAGAAGAGGA[C/T]GAGGAGCGTTTCCAGGGGTGGAAGC
Phenotype
MIM: 608306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SP8 PubMed Links

Gene Details

Gene
SP8
Gene Name
Sp8 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182700.5 352 Silent Mutation TCA,TCG S55S NP_874359.2
NM_198956.3 352 Silent Mutation TCA,TCG S37S NP_945194.1

View Full Product Details