Product Details

SNP ID
rs145939245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100107558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACGATGGGCTGCACGCTGGGGAC[A/G]GTGGTGGTGACGGGCGAAGTGGTGG
Phenotype
MIM: 602296 MIM: 600592 MIM: 602955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP4M1 PubMed Links

Gene Details

Gene
AP4M1
Gene Name
adaptor related protein complex 4 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004722.3 2244 Intron NP_004713.2
XM_005250689.4 2244 Intron XP_005250746.1
XM_005250690.4 2244 Intron XP_005250747.1
XM_006716175.3 2244 Intron XP_006716238.1
XM_011516685.2 2244 Intron XP_011514987.1
XM_017012790.1 2244 Intron XP_016868279.1
XM_017012791.1 2244 Intron XP_016868280.1
Gene
MCM7
Gene Name
minichromosome maintenance complex component 7
There are no transcripts associated with this gene.

Gene
TAF6
Gene Name
TATA-box binding protein associated factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190415.1 2244 Silent Mutation ACC,ACT T611T NP_001177344.1
NM_005641.3 2244 Silent Mutation ACC,ACT T574T NP_005632.1
NM_139315.2 2244 Silent Mutation ACC,ACT T574T NP_647476.1
XM_006716100.2 2244 Silent Mutation ACC,ACT T574T XP_006716163.1
XM_006716101.2 2244 Silent Mutation ACC,ACT T564T XP_006716164.1
XM_006716102.2 2244 Silent Mutation ACC,ACT T564T XP_006716165.1
XM_011516542.2 2244 Silent Mutation ACC,ACT T564T XP_011514844.1
XM_011516543.2 2244 Silent Mutation ACC,ACT T564T XP_011514845.1
XM_017012568.1 2244 Silent Mutation ACC,ACT T564T XP_016868057.1

View Full Product Details